Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHEMetrics™ by AquaPhoenix Vacu-vials Detergents Refill Kit, Use with I-2017 SAM Photometer.
SDP

Catalog No. 13299703
Change view
Click to view available options
Range:
0 to 2.5 ppm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Range
13-299-703 0 to 2.5 ppm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-299-703 Supplier CHEMetrics™ by AquaPhoenix Supplier No. R9423
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Instrumental refill kit containing double-point ampoules, test tubes, a dropper bottle, a tip-breaking tool, and instructions. Designed for use with the I-2017 SAM Photometer.

  • Methylene blue active substances (MBAS) bring about the transfer of methylene blue, a cationic dye, from an aqueous solution into an immiscible organic solvent.
  • This occurs through ion pair formation by the MBAS anion and the methylene blue cation. The intensity of the blue color in the organic solvent is directly related to the concentration of MBAS in the sample. Anionic surfactants commonly used in commercially available detergent formulations are prominent methylene blue active substances and are strongly responsive to this chemistry. Test results are expressed as ppm (mg/L) linear alkylbenzene sulfonate (LAS)

Specifications

pH Range 7
Volume (Metric) 0.5 mL
For Use With (Equipment) I-2017 SAM Photometer
No. of Tests 20
Range 0 to 2.5 ppm
Type Detergents Refill Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.