Learn More
CHEMetrics™ by AquaPhoenix Vacu-vials Iron Water Quality Instrumental Test Kit, Phenanthroline (Total and Ferrous)
SDP
Catalog No. 13299608
Change view
Click to view available options
No. of Tests:
30
Catalog No. No. of Tests
13-299-608 30
1 options
Catalog No. 13-299-608 Supplier CHEMetrics™ by AquaPhoenix Supplier No. K6203
Add to Cart
Add to Cart
Instrumental iron test kit featuring Vacu-vials ampoule technology for total and ferrous iron analysis. Compatible with most photometers and spectrophotometers.
  • Kit K-6203 comes in a cardboard box and contains everything needed to perform 30 tests: thirty ampoules, Activator Solution, 25 mL sample cup, ampoule blank, and instructions.
Specifications
Analytes Iron
pH Range 7
Volume (Metric) 0.5 mL
No. of Tests 30
Range 0 to 6.00 ppm
Type Water Quality Instrumental Test Kit