Learn More
CHEMetrics™ by AquaPhoenix Vacu-vials Molybdate Water Quality Instrumental Test Kit
SDP
Catalog No. 13299692
Change view
Click to view available options
No. of Tests:
30
Catalog No. No. of Tests
13-299-692 30
1 options
Catalog No. 13-299-692 Supplier CHEMetrics™ by AquaPhoenix Supplier No. K6703
Add to Cart
Add to Cart
Instrumental molybdate test kit supplied in a cardboard box with components for 30 tests. Requires a compatible photometer or spectrophotometer for analysis.
  • Kit comes in a cardboard box and contains everything needed to perform 30 tests: thirty ampoules, 25 mL sample cup, ampoule blank, and instructions.
Specifications
Analytes Molybdate
pH Range 7
Volume (Metric) 0.5 mL
No. of Tests 30
Range 0 to 25.0 ppm
Type Water Quality Instrumental Test Kit