Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHEMetrics™ by AquaPhoenix Vacu-vials Nitrate Water Quality Instrumental Test Kit, 0 to 1.5 ppm as N
SDP

Catalog No. 13299649
Change view
Click to view available options
No. of Tests:
30
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Tests
13-299-649 30
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-299-649 Supplier CHEMetrics™ by AquaPhoenix Supplier No. K6973
Add to Cart
Add to Cart

Instrumental nitrate test kit featuring self-filling Vacu-vials ampoule technology. The 13 mm ampoules are compatible with most photometers and spectrophotometers.

  • K-6973 Nitrate Vacu-vials test kit is used to measure the concentration of nitrate in water with zinc reduction.
  • It comes in a cardboard box and contains everything needed to perform 30 tests: thirty ampoules, Acidifier Solution, Buffer Solution, Zinc, reaction tube with lid, 25 mL sample cup, ampoule blank, and instructions.

Specifications

Analytes Nitrate
pH Range 7
Volume (Metric) 0.5 mL
No. of Tests 30
Range 0 to 1.5 ppm
Type Water Quality Instrumental Test Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.