Learn More
CHEMetrics Vacu-vials Test Kit, Chlorine 2
SDP
Catalog No. 13299525
Change view
Click to view available options
For Use With (Equipment):
V-2000 Photometer, Spectrophotometer, SAM Photometer
Catalog No. For Use With (Equipment)
13-299-525 V-2000 Photometer, Spectrophotometer, SAM Photometer
1 options
Catalog No. 13-299-525 Supplier CHEMetrics Supplier No. K2513
Add to Cart
Add to Cart
Use with any CHEMetrics Direct-Readout Photometer or spectrophotometer that accepts a 13 mm cell
Snap-and-read Vacu-vials self-filling ampules are the fastest, easiest reagent systems available for precision colorimetric determination of water/wastewater
  • Each vacuum-sealed ampule contains a unit dose of premeasured reagent
  • Simply immerse the ampule in the water sample, snap its tip, and an analytically correct amount of sample is pulled inside and mixed
  • Sample is now ready to be analyzed by your spectrophotometer (instrument must accept a 13 mm cell directly or with an adapter)
  • Direct readouts in concentration may be obtained by using CHEMetrics V-2000 Photometer or by preprogramming your spectrophotometer using standard solutions or the data pairs provided in the Vacu-vials instructions
  • For immediate results in your spectrophotometer without programming, use supplied equation or calibration tables to convert from absorbance or transmittance to concentration units
Specifications
Analytes Free and Total Chlorine 2
For Use With (Equipment) V-2000 Photometer, Spectrophotometer, SAM Photometer
Includes Cardboard box, 30 ampoules, activator solution, 25 mL sample cup, ampoule blank, instructions
No. of Tests 30
Range 0 to 5 ppm
Test Method DPD (free and total)
Test Time 1 min.
Type Vacu-vials Instrumental Test Kit