Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHEMetrics Vacu-vials Test Kit, Ammonia 1
SDP

Catalog No. 13299302
Change view
Click to view available options
For Use With (Equipment):
V-2000 Photometer or Spectrophotometer
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
13-299-302 V-2000 Photometer or Spectrophotometer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Contains mercury. Dispose according to local, state or federal laws.
Catalog No. 13-299-302 Supplier CHEMetrics Supplier No. K1503TAYLOR/DEL

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Contains mercury. Dispose according to local, state or federal laws.

Use with any CHEMetrics Direct-Readout Photometer or spectrophotometer that accepts a 13 mm cell

Snap-and-read Vacu-vials self-filling ampules are the fastest, easiest reagent systems available for precision colorimetric determination of water/wastewater

  • Each vacuum-sealed ampule contains a unit dose of premeasured reagent
  • Simply immerse the ampule in the water sample, snap its tip, and an analytically correct amount of sample is pulled inside and mixed
  • Sample is now ready to be analyzed by your spectrophotometer (instrument must accept a 13 mm cell directly or with an adapter)
  • Direct readouts in concentration may be obtained by using CHEMetrics V-2000 Photometer or by preprogramming your spectrophotometer using standard solutions or the data pairs provided in the Vacu-vials instructions
  • For immediate results in your spectrophotometer without programming, use supplied equation or calibration tables to convert from absorbance or transmittance to concentration units

Specifications

Analytes Ammonia 1
For Use With (Equipment) V-2000 Photometer or Spectrophotometer
Includes Cardboard box, 30 ampoules, stabilizer solution, 25 mL sample cup, ampoule blank, instructions, mercury (dispose according to local, state or federal laws)
No. of Tests 30
Range 0 to 7 ppm
Test Method Direct nesslerization
Test Time 2 min.
Type Vacu-vials Instrumental Test Kit
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.