Learn More
Greiner Bio-One™ VACUETTE™ Blood Culture Holder
Catalog No. 22040220
Change view
Click to view available options
Sterility:
Non-sterile
Catalog No. Sterility
22-040-220 Non-sterile
1 options
This item is not returnable. View return policy
Catalog No. 22-040-220 Supplier Greiner Bio-One™ Supplier No. 450181
Add to Cart
Add to Cart
This item is not returnable. View return policy
Designed for filling commonly used blood culture bottles and blood Collection Tubes.
  • Compatible with VACUETTE™ Blood Collection Needles.
  • Not made with natural rubber latex.
  • Non-sterile
  • Single-use device
TRUSTED_SUSTAINABILITY
Specifications
Material Polypropylene
Autoclavable Not Autoclavable
Color Transparent
Product Type Blood Culture Holder
Disposable Single-use
Latex Free Yes
Sterility Non-sterile