Learn More
Greiner Bio-One VACUETTE High Altitude Coagulation Tubes with Sodium Citrate Solution
Shop All Greiner Bio-One ProductsDescription
- Greiner Bio-One developed the VACUETTE Coagulation Tube, known as a 'sandwich' tube due to its patented double-wall technology, in 1996.
- The inner tube is made of polypropylene (PP) and prevents the evaporation of the sodium citrate additive to maintain the correct blood-to-additive ratio.
- The outer tube is made of polyethylene terephthalate (PET), which maintains the vacuum for maximum shelf life.
- VACUETTE Blood Collection Tubes are evacuated for easy, safe sample collection.
- The amount of vacuum in each tube corresponds to the tubes blood volume specification and helps ensure the correct blood-to-additive ratio.
- Injection molded from polyethylene terephthalate (PET) plastic, VACUETTE Blood Collection Tubes are highly transparent and virtually unbreakable.
- VACUETTE Serum tubes include a clot-activating additive micronized silica particles that is spray-dried on the interior tube wall for even disbursement and optimal blood/additive mixing.
- Optimal fill marks on every tube label indicate the proper draw volume for correct blood-to-additive ratios (within ±10 of the stated nominal volume in accordance with ISO and CLSI).
- Coagulation tube labels feature an arrow-shaped marker indicating the fill range where sample volume will yield the necessary 9:1 blood-to-additive ratio.
- Featuring blue VACUETTE SAFETY Caps, designed to minimize the spraying of aerosols during decapping and color-coded to aid first-glance tube recognition.
- The VACUETTE SAFETY Cap used on coagulation tubes is designed with a two-step stopper to seal both the inner and outer tubes for a longer shelf life.
- Sterile and latex-free.
Specifications
Specifications
| Color | Transparent |
| Product Type | Coagulation Tube |
| Additive | Sodium Citrate 3.2% |
| Material | PET |
| Sterility | Sterile |
| Closure Type | Non-ridged Pull Cap |
| Capacity (Metric) | 3 mL |
| Diameter (Metric) | 13 mm |
| Closure Color | Blue |
| Tube Size | 13 x 75 mm |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.