Learn More
Greiner Bio-One VACUETTE™ Urine Complete Set (Urine CCM / Urine STB Set with Cup and Towelette)
Shop All Greiner Bio-One ProductsDescription
The VACUETTE™ Urine Complete Set (Urine CCM / Urine STB Set with Cup and Towelette) includes:
- VACUETTE™ Urine CCM Tube for bacterial and yeast culture
- VACUETTE™ Urine STB Tube for urinalysis and sedimentation
- Urine Cup with Integrated Transfer Device for clean transfer of urine samples directly into VACUETTE™ Urine Tubes
- Castile soap towelette
Key benefits:
- Convenience: All you need, ready to use in a single easy-to-open pack
- Turnaround-time flexibility: VACUETTE™ Urine CCM tubes stabilize bacterial counts for up to 48 hours at room temperature while VACUETTE™ Urine STB tubes stabilize key parameters for urinalysis up to 72 hours unrefrigerated
Specifications
Specifications
| Product Type | Urine Specimen Collection Kit |
| Disposable | Single-use |
| Graduated | No |
| Material | Plastic |
| Sterility | Non-sterile |
| Color | Yellow |
| For Use With (Application) | Urine Microbiology, Clinical Chemistry, Sedimentation |
| Latex Free | Yes |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.