Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One VACUETTE™ Urine Complete Set (Urine CCM / Z Urine No Additive Set with Transfer Device and Towelette)

Catalog No. 21403461
Change view
Click to view available options
Sterility:
Non-sterile
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility
21-403-461 Non-sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-403-461 Supplier Greiner Bio-One Supplier No. 453049
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Designed to streamline collection and standardize workflows. These ready-to-use sets include all components needed for routine urine testing in an easy-open pouch. Available in several configurations to suit your facility's workflow.

The VACUETTE™ Urine Complete Set (Urine CCM / Z Urine No Additive Set with Transfer Device and Towelette) includes:

  • VACUETTE™ Urine CCM Tube for bacterial and yeast culture
  • VACUETTE™ Z Urine No Additive Tube for urinalysis and sedimentation
  • Urine Transfer Device for clean transfer of urine samples into VACUETTE™ Urine Tubes
  • Castile soap towelette
Key benefits:
  • Convenience: All you need, ready to use in a single easy-to-open pack
  • Turnaround-time flexibility: VACUETTE™ Urine CCM tubes stabilize bacterial counts for up to 48 hours at room temperature
TRUSTED_SUSTAINABILITY

Specifications

Product Type Urine Specimen Collection Kit
Disposable Single-use
Graduated No
Material Plastic
Sterility Non-sterile
Color Yellow
For Use With (Application) Urine Microbiology, Clinical Chemistry, Sedimentation
Latex Free Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.