Learn More
Electron Microscopy Sciences Vacule Caps-Solid Top, No Liner
SDP
Catalog No. 501898933
Encompass_Preferred
Heavy wall construction, designed for lyophilization and freeze-drying. Manufactured from Wheaton 33 low extractable borosilicate glass (meets ASTM Type I Class and USP Type I Standards). Accessory for 5 and 10 ml Vacule Vials.
Catalog No. 50-189-8933 Supplier Electron Microscopy Sciences Supplier No. 6030455
Heavy wall construction, designed for lyophilization and freeze-drying. Manufactured from Wheaton 33 low extractable borosilicate glass (meets ASTM Type I Class and USP Type I Standards). Accessory for 5 and 10 ml Vacule Vials.