Learn More
Electron Microscopy Sciences Vacule Red Stoppers
Supplier: Electron Microscopy Sciences 6030450
Heavy wall construction, designed for lyophilization and freeze-drying. Manufactured from Wheaton 33 low extractable borosilicate glass (meets ASTM Type I Class and USP Type I Standards). Accessory for 5 and 10 ml Vacule Vials.