Learn More
Chemglass Life Sciences Vacuum Gauge, Analog, Panel Mount, Complete
SDP
Catalog No. AF030101
Encompass_Preferred
Vacuum Gauge, Analog, Panel Mount, Complete. Economical thermocouple gauge has fast response and high stability. Gauge is supplied either in a bench top case with handle or panel mount style. Gauge utilizes a 8-pin Hastings-Raydist DV-6M thermocouple gauge tube having a 10ft cord. Gauge tube is threaded with 1/8” male NPT for use with AF-0306 adapter.
Catalog No. AF030101 Supplier Chemglass Life Sciences Supplier No. AF030101
Vacuum Gauge, Analog, Panel Mount, Complete. Economical thermocouple gauge has fast response and high stability. Gauge is supplied either in a bench top case with handle or panel mount style. Gauge utilizes a 8-pin Hastings-Raydist DV-6M thermocouple gauge tube having a 10ft cord. Gauge tube is threaded with 1/8” male NPT for use with AF-0306 adapter.