Learn More
Medchemexpress LLC Vacuolin-1 | 351986-85-1 | MFCD00321410 | 98.5% | 577.42 | C26H24IN7O | 5 MG
SDP
Catalog No. 5000228872
Encompass_Preferred
Vacuolin-1 (CAS 351986-85-1) is a cell-permeable small molecule that inhibits PIKfyve and blocks Ca2+-dependent lysosomal exocytosis. It impairs lysosomal maturation, inhibits late-stage autophagy, and induces vacuole formation, making it useful for studying endosomal/lysosomal trafficking and membrane dynamics in cell biology research.
  • Potent, cell-permeable PIKfyve inhibitor.
  • Blocks Ca2+-dependent lysosomal exocytosis in cells.
  • Impairs lysosomal maturation and inhibits late-stage autophagy.
  • Induces vacuole formation and alters endosomal/lysosomal trafficking.
  • Provided at high purity for research applications.
Catalog No. 50-002-28872 Supplier Medchemexpress LLC Supplier No. HY1186305MG
Vacuolin-1 (CAS 351986-85-1) is a cell-permeable small molecule that inhibits PIKfyve and blocks Ca2+-dependent lysosomal exocytosis. It impairs lysosomal maturation, inhibits late-stage autophagy, and induces vacuole formation, making it useful for studying endosomal/lysosomal trafficking and membrane dynamics in cell biology research.
  • Potent, cell-permeable PIKfyve inhibitor.
  • Blocks Ca2+-dependent lysosomal exocytosis in cells.
  • Impairs lysosomal maturation and inhibits late-stage autophagy.
  • Induces vacuole formation and alters endosomal/lysosomal trafficking.
  • Provided at high purity for research applications.