Learn More
Medchemexpress LLC Vacuolin-1 | 351986-85-1 | MFCD00321410 | 98.5% | 577.42 g·mol^-1 | C26H24IN7O | 10 MG
SDP
Catalog No. 5000227573
Encompass_Preferred
Vacuolin-1 is a cell-permeable small-molecule inhibitor of lysosomal exocytosis and PIKfyve used to study lysosome function and autophagy pathways.
  • High purity suitable for in vitro research.
  • Cell-permeable and active in cellular assays.
  • Induces vacuolation of endolysosomal compartments.
  • Soluble in DMSO for preparation of concentrated stock solutions.
  • Stable when stored under recommended conditions.
Catalog No. 50-002-27573 Supplier Medchemexpress LLC Supplier No. HY11863010MG
Vacuolin-1 is a cell-permeable small-molecule inhibitor of lysosomal exocytosis and PIKfyve used to study lysosome function and autophagy pathways.
  • High purity suitable for in vitro research.
  • Cell-permeable and active in cellular assays.
  • Induces vacuolation of endolysosomal compartments.
  • Soluble in DMSO for preparation of concentrated stock solutions.
  • Stable when stored under recommended conditions.