Learn More
Selleck Chemical LLC Vacuolin-1-5mg
SDP
Catalog No. 502389829
Encompass_Preferred
Vacuolin-1 is a potent and cell-permeable inhibitor that blocks the Ca(2+)-dependent exocytosis of lysosomes and prevents the release of lysosomal content without affecting the process of resealing. Vacuolin-1 is also a potent and selective PIKfyve inhibitor, and inhibits autophagy by impairing lysosomal maturation via PIKfyve inhibition.
Catalog No. 50-238-9829 Supplier Selleck Chemical LLC Supplier No. S69125MG
Vacuolin-1 is a potent and cell-permeable inhibitor that blocks the Ca(2+)-dependent exocytosis of lysosomes and prevents the release of lysosomal content without affecting the process of resealing. Vacuolin-1 is also a potent and selective PIKfyve inhibitor, and inhibits autophagy by impairing lysosomal maturation via PIKfyve inhibition.