Learn More
Cytiva (Formerly Pall Lab) Vacushield™ Vent Device - 50 mm, hose barb (3/pkg)
Supplier:  Cytiva 4402
Encompass
Protects Valves and Pump Components from Damage Due to Liquids-Incorporates a hydrophobic PTFE membrane-Allows air and gases to pass through freely while blocking aqueous solutions and aerosol contaminants-Offers highly effective retention of bacteria with minimal restriction of pump performance This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. NC0466009