Learn More
Med Vet International BD Vacutainer Venous Blood Collection Tube, ACD Solution B Additive, 6 mL, Conventional Closure Glass Tube, 100/Box
Catalog No. 503043030
Encompass_Preferred
Features ACD Solution B, 1.0 mL (Contains: Trisodium Citrate 13.2g / L, Citric Acid 4.8 g / L, Dextrose 14.7 g / L)
Catalog No. 50-304-3030 Supplier Med Vet International Supplier No. 364816
Features ACD Solution B, 1.0 mL (Contains: Trisodium Citrate 13.2g / L, Citric Acid 4.8 g / L, Dextrose 14.7 g / L)