Learn More
Med Vet International BD Vacutainer Venous Blood Collection Tube, ACD Solution A Additive, 8.5 mL, Conventional Closure Glass Tube, 100/Box
Catalog No. 503044709
Encompass_Preferred
Features ACD Solution A, 1.5 ml (Contains: Trisodium Citrate 22.0 g/ L, Citric Acid 8.0 g/L, Dextrose 24.5 g/L) Blood banking / histocompatibilty / sterile vacuum collection for blood group determination
Catalog No. 50-304-4709 Supplier Med Vet International Supplier No. 364606
Features ACD Solution A, 1.5 ml (Contains: Trisodium Citrate 22.0 g/ L, Citric Acid 8.0 g/L, Dextrose 24.5 g/L) Blood banking / histocompatibilty / sterile vacuum collection for blood group determination