Learn More
BD Vacutainer™ Blood Collection Tube, Trace Element K2
Catalog No. B367777 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Capacity (Metric):
3 mL
Catalog No. Capacity (Metric)
B367777 3 mL
1 options
This item is not returnable. View return policy
Catalog No. B367777 Supplier BD Supplier No. 367777
Add to Cart
Add to Cart
This item is not returnable. View return policy
Vacutainer™ Blood Collection Tube for collection and transport of blood samples.
  • Trace Element K2 EDTA
  • 13 x 75 mm
  • 3.0 mL
  • Royal Blue Hemogard
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
Product Type Blood Collection Tube
Additive EDTA
Material Plastic
Sterility Non-sterile
Closure Type Cap
Capacity (Metric) 3 mL