Learn More
Med Vet International BD Vacutainer Quick Release Needle Holder, for Blood Collection, 20/Box
Catalog No. 503044861
Encompass_Preferred
Product Description Each box contains 20 single-use BD Vacutainer® Holders and adapters, suitable for both 13mm and 16mm tubes and Bactec™ blood culture bottles. These options are designed to support optimal infection control practices, providing high-quality and cost-efficient solutions.  The BD Vacutainer® One Use Holder is compatible with BD Vacutainer® Eclipse™ Blood Collection Needles, BD Vacutainer® Safety-Lok™ Blood Collection Sets, BD Vacutainer® Push Button Blood Collection Sets, and the BD Vacutainer® Multi Sample Luer Adapter.
Catalog No. 50-304-4861 Supplier Med Vet International Supplier No. 368872
Product Description Each box contains 20 single-use BD Vacutainer® Holders and adapters, suitable for both 13mm and 16mm tubes and Bactec™ blood culture bottles. These options are designed to support optimal infection control practices, providing high-quality and cost-efficient solutions.  The BD Vacutainer® One Use Holder is compatible with BD Vacutainer® Eclipse™ Blood Collection Needles, BD Vacutainer® Safety-Lok™ Blood Collection Sets, BD Vacutainer® Push Button Blood Collection Sets, and the BD Vacutainer® Multi Sample Luer Adapter.