Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Vacutainer™ Latex-Free Tourniquets

Catalog No. 026576 Shop All Waters Corporation Products
Change view
Click to view available options
Dimensions (L x W):
18 x 1 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
02-657-6 18 x 1 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 02-657-6 Supplier BD Supplier No. 367203
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Stretch, latex-free

  • Nonpinching
  • W x L: 1 × 18 in. (2.5 × 45.7cm)

TRUSTED_SUSTAINABILITY

Specifications

Latex Free Yes
Product Type Tourniquet
For Use With (Application) Perforated design allows for easy, one-at-a-time dispensing that facilitates a single-use policy
Length (Metric) 45.7 cm
Length (English) 18 in.
Width (English) 1 in.
Width (Metric) 2.5 cm
Dimensions (L x W) 18 x 1 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.