Learn More
Med Vet International BD Vacutainer Multiple Sample, Luer Adapter, Male Slip-Luer Fitting, Sterile, Holder, 100/Box
Catalog No. 503043031
Encompass_Preferred
Features The BD Vacutainer® Luer Adapter is a male slip-luer fitting opposing a multiple sample non-patient (NP) needle It is designed to be used with a BD Vacutainer® Holder (Not Included) Used for sample transfer
Catalog No. 50-304-3031 Supplier Med Vet International Supplier No. 367290
Features The BD Vacutainer® Luer Adapter is a male slip-luer fitting opposing a multiple sample non-patient (NP) needle It is designed to be used with a BD Vacutainer® Holder (Not Included) Used for sample transfer