Learn More
Med Vet International BD Vacutainer, Multi-sample Needles, 20G x 1in, Yellow, 100/Box
Catalog No. 503042916
Encompass_Preferred
The BD Vacutainer™ system is optimally designed and tested for controlling pre-analytical variability throughout the specimen collection and handling process. Sterile double-ended needle with a sleeve covered safety valve Advanced low-angle bevel design and silicone lubricant coating ensures gentle and smooth vein entry Draws blood directly from the vein into vacuum collection tubes to prevent exposure Can be used with multiple collection tubes after venipuncture Designed for use with BD Vacutainer™ tube holders and vacuum blood collection tubes Luer adapter (MPN 367300) available separately for use with a catheter or hypodermic needle
Catalog No. 50-304-2916 Supplier Med Vet International Supplier No. 360214
The BD Vacutainer™ system is optimally designed and tested for controlling pre-analytical variability throughout the specimen collection and handling process. Sterile double-ended needle with a sleeve covered safety valve Advanced low-angle bevel design and silicone lubricant coating ensures gentle and smooth vein entry Draws blood directly from the vein into vacuum collection tubes to prevent exposure Can be used with multiple collection tubes after venipuncture Designed for use with BD Vacutainer™ tube holders and vacuum blood collection tubes Luer adapter (MPN 367300) available separately for use with a catheter or hypodermic needle