Learn More
Med Vet International BD Vacutainer, PrecisionGlide Needles, 21G x 1in, Green, 100/Box
Catalog No. 503044712
Encompass_Preferred
Features 21 G x 1" Clear hub Polystyrene hub material  Threaded hub Regular needle tip Blood collection needle Thin needle wall Sterile B-level bevel DEHP-free Latex-free Disposable
Catalog No. 50-304-4712 Supplier Med Vet International Supplier No. 360212
Features 21 G x 1" Clear hub Polystyrene hub material  Threaded hub Regular needle tip Blood collection needle Thin needle wall Sterile B-level bevel DEHP-free Latex-free Disposable