Learn More
BD Vacutainer™ P800 Blood Collection System
Catalog No. 22011108 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Capacity (Metric):
2 mL
8.5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
22-011-108 8.5 mL
21-402-721 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-011-108 Supplier BD Supplier No. 366421
Add to Cart
Add to Cart
This item is not returnable. View return policy
BD™ P800 delivers enhanced accuracy in measuring plasma metabolic markers such as Glucagon-Like Peptide-1 (GLP-1), Glucagon, non-acyl Ghrelin, Gastric Inhibitory Polypeptide (GIP), and Oxyntomodulin (OXM) by stabilizing them at the point of collection.
WHY IT'S NEEDED
Labile metabolic peptides (e.g., GLP-1, GIP, glucagon, OXM, ghrelin) degrade rapidly in conventional EDTA plasma, reducing accuracy and reproducibility. Stabilizing intact analytes early minimizes preanalytical variability.
HOW IT WORKS
A standardized workflow and a specialized tube with proprietary protease, esterase, and DPP-IV inhibitors that act immediately at blood draw to halt enzymatic activity
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
Product Type Blood Collection Tube
Additive None
Material Plastic
Sterility Sterile
Closure Type Hemogard
Capacity (Metric) 8.5 mL