Learn More
BD Vacutainer™ Serum Separator (SST™) Blood Collection Tubes
Shop All BD Diagnostic Systems ProductsDescription
- Contains clot activator and polymer gel for serum separator tube (SST™) applications
- Designed with a 10 mL draw volume for blood specimen collection
- Features a 16 mm outer diameter and 16 x 125 mm tube size
- Red/gray color configuration supports visual identification of the tube
- Features a conventional closure design
- Supplied with a paper label for specimen identification
- Supplied by the case with 1,000 tubes per case
Specifications
Specifications
| Color | Red |
| Product Type | Blood Collection Tube |
| Additive | Clot Activator and Gel |
| Material | Plastic |
| Sterility | Sterile |
| Closure Type | Conventional Stopper |
| Capacity (Metric) | 10 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.