Learn More
Med Vet International BD Vacutainer SPC Plus Venous Blood Collection Tube, Clot Activator Additive, 6 mL, Hemogard™ Closure Plastic Tube, 100/Box
Catalog No. 503044729
Encompass_Preferred
Features BD Medical 368380 Vacutainer® Plus Plastic Blood Collection Tubes for Trace Element Testing Paper Label and Royal Blue Hemogard™ Closure These Vacutainer® Blood Collection Tubes are designed for Trace Element Testing in Blood 510(k) Cleared
Catalog No. 50-304-4729 Supplier Med Vet International Supplier No. 368380
Features BD Medical 368380 Vacutainer® Plus Plastic Blood Collection Tubes for Trace Element Testing Paper Label and Royal Blue Hemogard™ Closure These Vacutainer® Blood Collection Tubes are designed for Trace Element Testing in Blood 510(k) Cleared