Learn More
BD Vacutainer™ Venous Blood Collection Tubes: SST™ Serum Separation Tubes
Catalog No. 02683148 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Capacity (Metric):
8.5 mL
10 mL
13 x 75 mm
16 x 100 mm
Closure Color:
Red/Gray
Red/Green
Additive:
Polymer Gel/Silica Activator
Silica/Gel
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Closure Color Additive
02-683-148 16 x 100 mm Red/Green Silica/Gel
02-683-96 8.5 mL Red/Gray Polymer Gel/Silica Activator
02-683-98 10 mL Red/Gray Polymer Gel/Silica Activator
15-350-30 13 x 75 mm Red/Gray Polymer Gel/Silica Activator
02-683-95 16 x 100 mm Red/Gray Polymer Gel/Silica Activator
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 02-683-148 Supplier BD Supplier No. 368016
This item is not returnable. View return policy
Ideal for use in serum determinations in chemistry
  • Contain spray-coated silica and a polymer gel for serum separation
  • Provide an efficient means of serum sample preparation and help to improve laboratory workflow
  • Tubes marked as Best Practice Formulary meet demanding requirements of complex diagnostic instruments and current clinical guidelines
  • Available with gold conventional tube stopper or red/green Hemogard™ tube closure that prevents human contact with blood on the stopper or tube rim
Evacuated tubes simplify blood collection
  • Blood is drawn directly from the vein into the evacuated sterile collection tubes
  • Only one venipuncture is needed to draw multiple samples
  • Tubes can be changed while needle and plastic holder remain in place on patient's arm
  • Closures are color coded
Plastic Evacuated Tubes
  • Made of polyethylene terephthalate (PET)
  • Provide a safer work environment for all personnel by (1) reducing risk of tube breakage during handling; (2) eliminating breakage when tubes are centrifuged at higher speeds; and (3) eliminating breakage or chipping of tube rims when stopper is removed and reinserted
  • In addition to reducing potential injuries, fewer tube breakages result in less workflow interruption and fewer patient redraws, providing smoother day-to-day operation
Hemogard™ Stoppers
  • Safety shield designed for safer tube handling
  • Rubber stopper is recessed inside the plastic shield, so any drops left by a blood collection needle remain isolated from potential contact
  • Shield is designed to protect lab personnel from direct contact with blood on the stopper or around the outer rim of the tube, as well as from blood splattering upon opening the tube
TRUSTED_SUSTAINABILITY
Specifications
Additive Silica/Gel
Material Plastic
Closure Type Hemogard
Capacity (Metric) 16 x 100 mm
Closure Color Red/Green
For Use With (Application) For use in serum determinations in chemistry