Learn More
Med Vet International Vacutainer, Needle Holder, Single Use
Supplier:  Med Vet International 364815
Encompass_Preferred
The BD 364815 Vacutainer Needle Holder provides convenience and promotes safety. You can avoid the bother of storing and cleaning reusable holders with this one-use, non-stackable needle holder. It also reduces the hazards of contact with blood-borne pathogens compared to reusable holders. The Vacutainer Needle Holder is an affordable alternative whether you are placing a large or small order. The non-sterile holder is clear and latex free.
Catalog No. 50-209-3085
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
View Alternate Product