Learn More
Sigma Aldrich Fine Chemicals Biosciences Vacuubrand Chemistry vacuu1EA
Catalog No. 502908731
Encompass
Three-stage chemistry-design diaphragm pumps are an excellent solution for continuous oil-free pumping of corrosive gases and vapors and meet many difficult requirements. The three-stage construction provides the advantageous combination of high pumping speed and very low ultimate vacuum. All major parts in contact with pumped media are made of chemically resistant fluoroplastics.Well-proven PTFE sandwich diaphragms provide increased reliability and extended operating life. The MD 4C NT with gas ballast valve is optimally prepared for pumping condensable vapors due to the very good ultimate vacuum (even with gas ballast) as well as for high boiling solvents. The new NT-series features further improve performance data and superior vapor tolerance..nbspPump supplied completely mounted ready for use with manual.nbspOutstanding chemical resistance and superior vapor tolerance.nbspExceptionally high performance even at low vacuum.nbspExcellent ultimate vacuum even with gas ballast.nbspMa
Catalog No. 50-290-8731 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z6762331EA
Add to Cart
Add to Cart
Request a Quote
Three-stage chemistry-design diaphragm pumps are an excellent solution for continuous oil-free pumping of corrosive gases and vapors and meet many difficult requirements. The three-stage construction provides the advantageous combination of high pumping speed and very low ultimate vacuum. All major parts in contact with pumped media are made of chemically resistant fluoroplastics.Well-proven PTFE sandwich diaphragms provide increased reliability and extended operating life. The MD 4C NT with gas ballast valve is optimally prepared for pumping condensable vapors due to the very good ultimate vacuum (even with gas ballast) as well as for high boiling solvents. The new NT-series features further improve performance data and superior vapor tolerance..nbspPump supplied completely mounted ready for use with manual.nbspOutstanding chemical resistance and superior vapor tolerance.nbspExceptionally high performance even at low vacuum.nbspExcellent ultimate vacuum even with gas ballast.nbspMa