Learn More
Sigma Aldrich Fine Chemicals Biosciences Vacuubrand Chemistry vacuu1EA
Catalog No. 502908662
Encompass
Three-stage chemistry-design diaphragm pumps are an excellent SOLUTIONon for continuous oil-free pumping of corrosive gases and vapors and meet many difficult requirements. The three-stage construction provides the advantageous combination of high pumping speed and very low ultimate vacuum. All major parts in contact with pumped media are made of chemically resistant fluoroplastics.Well-proven PTFE sandwich diaphragms provide increased reliability and extended operating life. The MD 4C NT with gas ballast valve is optimally prepared for pumping condensable vapors due to the very good ultimate vacuum (even with gas ballast) as well as for high boiling solvents. The new NT-series features further improve performance data and superior vapor tolerance.Pump supplied completely mounted ready for use with manualOutstanding chemical resistance and superior vapor toleranceExceptionally high performance even at low vacuumExcellent ultimate vacuum even with gas ballastMa
Catalog No. 50-290-8662 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z6762761EA
Three-stage chemistry-design diaphragm pumps are an excellent SOLUTIONon for continuous oil-free pumping of corrosive gases and vapors and meet many difficult requirements. The three-stage construction provides the advantageous combination of high pumping speed and very low ultimate vacuum. All major parts in contact with pumped media are made of chemically resistant fluoroplastics.Well-proven PTFE sandwich diaphragms provide increased reliability and extended operating life. The MD 4C NT with gas ballast valve is optimally prepared for pumping condensable vapors due to the very good ultimate vacuum (even with gas ballast) as well as for high boiling solvents. The new NT-series features further improve performance data and superior vapor tolerance.Pump supplied completely mounted ready for use with manualOutstanding chemical resistance and superior vapor toleranceExceptionally high performance even at low vacuumExcellent ultimate vacuum even with gas ballastMa