Learn More
Med Vet International VacuuForm Surgical Positioner - Small, J1582, Each
Catalog No. 5000583047
Encompass_Preferred
The VacuuForm positioner has small microbeads that when the air is removed by a vacuum hand pump, they are compressed into a desired rigid shape that maintains the patient in a desired position. Use for radiology, ultrasound or surgery, and safe for MRIs. Just open valve to release vacuum and the positioner returns to soft, pliable feel. • 100% Polyurethane blue material • Very lightweight • Washable • Soft but strong material • Radiolucent • Hand pump w/flexible hose connector included. VacuuForm Surgical Positioner - Small. Dimensions (WxL) 16 x 32(in) 40 x 60 (cm
Catalog No. 50-005-83047 Supplier Med Vet International Supplier No. JOR1582
The VacuuForm positioner has small microbeads that when the air is removed by a vacuum hand pump, they are compressed into a desired rigid shape that maintains the patient in a desired position. Use for radiology, ultrasound or surgery, and safe for MRIs. Just open valve to release vacuum and the positioner returns to soft, pliable feel. • 100% Polyurethane blue material • Very lightweight • Washable • Soft but strong material • Radiolucent • Hand pump w/flexible hose connector included. VacuuForm Surgical Positioner - Small. Dimensions (WxL) 16 x 32(in) 40 x 60 (cm