Learn More
Waters Corp Vacuum Manifold, Semi-automated, SPE, Well Plates
Catalog No. 502391526 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This vacuum manifold is a semi-automated solution for Solid Phase Extraction (SPE) in well plates. It connects with OneLab for automated pump control and features a machine-readable design to enable SPE methods with the Andrew+ robot.
  • Semi-automated solution for Solid Phase Extraction (SPE) in well plates.
  • Connects with OneLab for automated pump control, including on/off functionality.
  • Features a machine-readable design for SPE methods.
  • Compatible with the Andrew+ robot.
Catalog No. 50-239-1526 Supplier Waters Corp Supplier No. 176004579
Add to Cart
Add to Cart
Request a Quote
This vacuum manifold is a semi-automated solution for Solid Phase Extraction (SPE) in well plates. It connects with OneLab for automated pump control and features a machine-readable design to enable SPE methods with the Andrew+ robot.
  • Semi-automated solution for Solid Phase Extraction (SPE) in well plates.
  • Connects with OneLab for automated pump control, including on/off functionality.
  • Features a machine-readable design for SPE methods.
  • Compatible with the Andrew+ robot.