Learn More
Sigma Aldrich Fine Chemicals Biosciences Vacuum accessories for Buc1EA
Catalog No. 502909016
Encompass
This valve comes with a Woulff bottle and an attachment for Rotavapors. Not to be used with V700/710 pump in stand alone systems.
Catalog No. 50-290-9016 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z5650081EA
This valve comes with a Woulff bottle and an attachment for Rotavapors. Not to be used with V700/710 pump in stand alone systems.