Learn More
Electron Microscopy Sciences Vacuum Bell Jar & Plate
SDP
Catalog No. 50930948
Encompass_Preferred
Good for vacuum drying and degassing procedures. The chamber is made from a clear polycarbonate allowing for full visibility. Its polypropylene stopcock has a "PTFE" plug and it accepts 1/4" (6.4 mm) I.D. tubing. The chamber measures: 7 7/8" (20cm) I.D.; inside maximum height is 4 3/8" (11cm); plate is 1 3/4" (4.4cm) H x 9 1/8" (23.2cm) in diameter.
Catalog No. 50-930-948 Supplier Electron Microscopy Sciences Supplier No. 71236
Good for vacuum drying and degassing procedures. The chamber is made from a clear polycarbonate allowing for full visibility. Its polypropylene stopcock has a "PTFE" plug and it accepts 1/4" (6.4 mm) I.D. tubing. The chamber measures: 7 7/8" (20cm) I.D.; inside maximum height is 4 3/8" (11cm); plate is 1 3/4" (4.4cm) H x 9 1/8" (23.2cm) in diameter.