Learn More
Bellco Glass, Inc. Vacuum Borosilicate Glass Filtration Assembly 90MM, 5000 ml flask 1000 ml funnel
SDP
Catalog No. 502549941
Encompass_Preferred
Borosil Filtration All Glass Filter Holder Assembly, 90mm, 1/EABorosil Filtration Assemblies are used for the filtration of HPLC solvents and fluid mobile phase solutions and buffers. Each assembly is made of 100% Borosilicate glass with a fritted glass filter base. The base design has an integral vacuum connection located above the filtrate drip to prevent contamination of the vacuum line. Comes with an anodized aluminum spring clamp. The filter size is 90mm.SpecificationsMaterial: 3.3 Borosilicate GlassColor: ClearFiltration Base Size: 90mmClass/ Quality Grade: Type IQuantity Per Case: 1
Catalog No. 50-254-9941 Supplier Bellco Glass, Inc. Supplier No. 452800821
Borosil Filtration All Glass Filter Holder Assembly, 90mm, 1/EABorosil Filtration Assemblies are used for the filtration of HPLC solvents and fluid mobile phase solutions and buffers. Each assembly is made of 100% Borosilicate glass with a fritted glass filter base. The base design has an integral vacuum connection located above the filtrate drip to prevent contamination of the vacuum line. Comes with an anodized aluminum spring clamp. The filter size is 90mm.SpecificationsMaterial: 3.3 Borosilicate GlassColor: ClearFiltration Base Size: 90mmClass/ Quality Grade: Type IQuantity Per Case: 1