Learn More
Electron Microscopy Sciences Heavy Duty Vacuum Bottle w/screw cap, Material: HDPE. Cap: 83 mm., 6/PK
SDP
Catalog No. 502090869
Encompass_Preferred
These all new Heavy Duty Vacuum Bottles have thick walls to withstand the forces of repeated vacuum applications in the laboratory. Designed for liquid aspiration and transfer operations, the bottles come complete with a polypropylene cap and TPE gasket for leakproof sealing. When combined with the matching aseptic transfer caps, these bottles can be used for safe, sterile transfer of reagents, media and solutions required in biotechnology, cell culture and other laboratories. Material: HDPE. Cap: 83 mm .
Catalog No. 50-209-0869 Supplier Electron Microscopy Sciences Supplier No. 6426208
These all new Heavy Duty Vacuum Bottles have thick walls to withstand the forces of repeated vacuum applications in the laboratory. Designed for liquid aspiration and transfer operations, the bottles come complete with a polypropylene cap and TPE gasket for leakproof sealing. When combined with the matching aseptic transfer caps, these bottles can be used for safe, sterile transfer of reagents, media and solutions required in biotechnology, cell culture and other laboratories. Material: HDPE. Cap: 83 mm .