Learn More
Waters Corp Waters Vacuum Box Gasket Kit, for 96-Well Extraction Plate Manifold
Catalog No. 50781818 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This kit is designed for use with 96-well extraction plate manifolds. It provides essential components for maintenance and optimal performance.
  • Designed for 96-well extraction plate manifolds.
  • Includes foam top gaskets.
  • Includes orange O-rings.
Catalog No. 50-781-818 Supplier Waters Corp Supplier No. 186003522
This kit is designed for use with 96-well extraction plate manifolds. It provides essential components for maintenance and optimal performance.
  • Designed for 96-well extraction plate manifolds.
  • Includes foam top gaskets.
  • Includes orange O-rings.