Learn More
Grainger Vacuum Breaker Repair Kit Coyne Delaney
Catalog No. 502753356
Encompass_Preferred
The KISSLER Vacuum Breaker Repair Kit is designed to service the KISSLER Model 609 and 909 vacuum breakers. The kit includes the necessary parts for rebuilding the vacuum breaker, such as the check stop assembly, check valve, critical level float, air inlet valve, and associated gaskets and O-rings. The vacuum breaker protects against backsiphonage and backpressure of contaminated water entering the potable water system. This repair kit allows you to rebuild the internal components of the vacuum breaker and extend its service life. The kit fits 1/2", 3/4", and 1" vacuum breakers.
Catalog No. 50-275-3356 Supplier Grainger Supplier No. 14U335
The KISSLER Vacuum Breaker Repair Kit is designed to service the KISSLER Model 609 and 909 vacuum breakers. The kit includes the necessary parts for rebuilding the vacuum breaker, such as the check stop assembly, check valve, critical level float, air inlet valve, and associated gaskets and O-rings. The vacuum breaker protects against backsiphonage and backpressure of contaminated water entering the potable water system. This repair kit allows you to rebuild the internal components of the vacuum breaker and extend its service life. The kit fits 1/2", 3/4", and 1" vacuum breakers.