Learn More
BUCHI Vacuum Control for L-250
SDP
Catalog No. 01670498 Shop All BUCHI Products
Change view
Click to view available options
Style:
Advanced Vacuum Control
Ultimate Vacuum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Style
01-670-498 Advanced Vacuum Control
01-670-499 Ultimate Vacuum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-670-498 Supplier BUCHI Supplier No. 11080162
Add to Cart
Add to Cart
Request a Quote
The Lyovapor™ sensors accurately track temperature and/or pressure throughout the freeze-drying process, ensuring optimized operations and facilitating accurate endpoint determination using capacitive, Pirani pressure gauges, and sample temperature probes.
  • Chose from ultimate vacuum (no control) or advanced vacuum control for process regulation and method development
Specifications
Type Benchtop Freeze Dryer
Capacity (Metric) 4.5 L
No. of Shelves 0
Temperature Range (Metric) Ambient to -105°C
Material Stainless Steel
Style Advanced Vacuum Control
Plug Type North America
Voltage 220/240 V