Learn More
Electron Microscopy Sciences Handi-Vac with 1/2" Static Dissipative Vacuum Cup
SDP
Catalog No. 501929311
Encompass_Preferred
The Handi-Vac-2 has an improved tip designed for better lifting capacity utilizing larger, non-marking vacuum cups. This new tip also enhances functionality and improves accuracy when picking and placing parts. One static-dissipative cup included. The entire vacuum tool is ESD-safe.
Catalog No. 50-192-9311 Supplier Electron Microscopy Sciences Supplier No. 7192102
The Handi-Vac-2 has an improved tip designed for better lifting capacity utilizing larger, non-marking vacuum cups. This new tip also enhances functionality and improves accuracy when picking and placing parts. One static-dissipative cup included. The entire vacuum tool is ESD-safe.