Learn More
Research Products International Corp Lab Companion Vacuum Desiccator, UV Blocking Amber, 10 Liter
SDP
Catalog No. 501367912
Encompass
These high-quality vacuum desiccators stay gastight allowing reliable dry storage or experiments over extended periods. Able to maintain 29in.-Hg vacuum for over 72 hours at room temperature, they provide vacuum integrity resulting in lower maintenance and greater peace of mind.
  • Viton 3-way valve offers consistent and uniform vacuum draw, vacuum release, or gas exchange
  • Vacuum level checking is simplified with a built-in analog vacuum gauge at the top of the unit
  • High-quality silicone O-ring and specially designed locking ring allow outstanding vacuum capability with virtually no leakage
  • strong>WARNING  This product can expose you to chemicals including Lead and Bisphenol-A, which is known to the State of California to cause cancer or birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
Catalog No. 50-136-7912 Supplier Research Products International Corp Supplier No. 420051A
These high-quality vacuum desiccators stay gastight allowing reliable dry storage or experiments over extended periods. Able to maintain 29in.-Hg vacuum for over 72 hours at room temperature, they provide vacuum integrity resulting in lower maintenance and greater peace of mind.
  • Viton 3-way valve offers consistent and uniform vacuum draw, vacuum release, or gas exchange
  • Vacuum level checking is simplified with a built-in analog vacuum gauge at the top of the unit
  • High-quality silicone O-ring and specially designed locking ring allow outstanding vacuum capability with virtually no leakage
  • strong>WARNING  This product can expose you to chemicals including Lead and Bisphenol-A, which is known to the State of California to cause cancer or birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.