Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DWK Life Sciences™ Kimble™ Vacuum Desiccator Lid

Replacement lid for 150mm, 20mm or 250mm inner diameter Vacuum Desiccator with Collar
Supplier: DWK Life Sciences™ 31252200
Description
Material: Soda lime glass
Includes top knob

Specifications
Vacuum Desiccator Lid | |
Soda Lime Glass | |
312200-200: 200mm inner diameter Vacuum Desiccator with Collar | |
Top knob |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction