Learn More
Foxx Life Sciences Abdos Vacuum Desiccator with a clear Polycarbonate (PC) top and a white Polypropylene (PP) Plate (Diameter 150mm) 1/EA
GREENER_CHOICE
Catalog No. NC1818243
Encompass_Preferred
Vacuum desiccators are molded with high-quality plastics which are light and easy to handle. Desiccators are enclosures containing desiccants for preserving moisture sensitive products and the vacuum chamber accelerates drying. The Polycarbonate (PC) top is clear for easy viewing while the bottom part of the desiccators are white Polypropylene (PP) and come with a thick white perforated disc. Silicone O-ring assures tight seal down to 0°C without using grease. •Polypropylene stopcock with PTFEFoxx Life Sciences is proud to represent high-quality Abdos plasticware and plastic laboratory products. For downloadable Certificates please visit: http://www.abdoslifesciences.com/Global/certificates.html.
Catalog No. NC1818243 Supplier Foxx Life Sciences Supplier No. E11605
Vacuum desiccators are molded with high-quality plastics which are light and easy to handle. Desiccators are enclosures containing desiccants for preserving moisture sensitive products and the vacuum chamber accelerates drying. The Polycarbonate (PC) top is clear for easy viewing while the bottom part of the desiccators are white Polypropylene (PP) and come with a thick white perforated disc. Silicone O-ring assures tight seal down to 0°C without using grease. •Polypropylene stopcock with PTFEFoxx Life Sciences is proud to represent high-quality Abdos plasticware and plastic laboratory products. For downloadable Certificates please visit: http://www.abdoslifesciences.com/Global/certificates.html.