Learn More
Electron Microscopy Sciences Multifunctional Vacuum Desiccators, 1/EA
SDP
Catalog No. 502091026
Encompass
The vacuum desiccator stores grids and or mounts under vacuum and eliminates dust particles and airborne contamination and prevents oxidation. The base desiccator pumps down to 10-3 mbar and can hold vacuum for up to one year. The versatile holder for the storage as well as shipping of samples.< br /> The unit is made from Red Anodized aluminum and it measures 76 mm x 35 mm (3" x 1.4") and has a standard vacuum connection (¼") (6.35 mm) barb fitting.< br /> The desiccators may be used with any laboratory pump including and the EMS 94588 series of pumps. Comes complete with an acrylic lid to enable visualization of samples.
Catalog No. 50-209-1026 Supplier Electron Microscopy Sciences Supplier No. 7387901
Add to Cart
Add to Cart
The vacuum desiccator stores grids and or mounts under vacuum and eliminates dust particles and airborne contamination and prevents oxidation. The base desiccator pumps down to 10-3 mbar and can hold vacuum for up to one year. The versatile holder for the storage as well as shipping of samples.< br /> The unit is made from Red Anodized aluminum and it measures 76 mm x 35 mm (3" x 1.4") and has a standard vacuum connection (¼") (6.35 mm) barb fitting.< br /> The desiccators may be used with any laboratory pump including and the EMS 94588 series of pumps. Comes complete with an acrylic lid to enable visualization of samples.