Learn More
Foxx Life Sciences Borosil® Glass Vacuum Desiccator with Porcelain Plate and Borosilicate Lid with Glass Stopcock and PTFE Spindle, Extra Small (XS), 100 mm Diameter, 1/EA
Catalog No. 501720182
Encompass_Preferred
From streamlined contours to high-quality glass stopcocks, Borosil® Vacuum Desiccators reflect great care and attention to detail. Borosil® Desiccators are perfect for drying, and for storage of moisture-sensitive substances. Compatible with any common desiccant. Porcelain plate, ground glass flanges. Rated to 9.6 PSI vacuum when properly cleaned and greased. Available from 100mm (XS) to 300mm (XL). Also available in Light-Blocking Amber. In collaboration with Borosil Glass Works, Foxx Life Sciences is proud to bring the outstanding quality of Borosil® Type 3.3 Borosilicate glassware to North America for the first time. Manufactured from the same USP Type 1 borosilicate glass that North American scientists have come to trust, Borosil® Laboratory Glassware offers unparalleled mechanical strength, clarity, and reliability at competitive prices. And thanks to a unique annealing process, Borosil® Glassware is stress-free and incredibly resistant to thermal shock.
Catalog No. 50-172-0182 Supplier Foxx Life Sciences Supplier No. 3083041
From streamlined contours to high-quality glass stopcocks, Borosil® Vacuum Desiccators reflect great care and attention to detail. Borosil® Desiccators are perfect for drying, and for storage of moisture-sensitive substances. Compatible with any common desiccant. Porcelain plate, ground glass flanges. Rated to 9.6 PSI vacuum when properly cleaned and greased. Available from 100mm (XS) to 300mm (XL). Also available in Light-Blocking Amber. In collaboration with Borosil Glass Works, Foxx Life Sciences is proud to bring the outstanding quality of Borosil® Type 3.3 Borosilicate glassware to North America for the first time. Manufactured from the same USP Type 1 borosilicate glass that North American scientists have come to trust, Borosil® Laboratory Glassware offers unparalleled mechanical strength, clarity, and reliability at competitive prices. And thanks to a unique annealing process, Borosil® Glassware is stress-free and incredibly resistant to thermal shock.