Learn More
Foxx Life Sciences Borosil Glass Vacuum Desiccator with Porcelain Plate and Borosilicate Lid with Glass Stopcock and PTFE Spindle, Large (L), 300mm Diameter, 1/EA
Catalog No. 501542839
Encompass_Preferred
From streamlined contours to high-quality glass stopcocks, Borosil Vacuum Desiccators reflect great care and attention to detail. Borosil Desiccators are perfect for drying, and for storage of moisture-sensitive substances. Compatible with any common desiccant. Porcelain plate, ground glass flanges. Rated to 9.6 PSI vacuum when properly cleaned and greased. Avail. from 100mm (XS) to 300mm (XL). Also Avail. in Light-Blocking Amber. In collaboration with Borosil Glass Works, Foxx Life Sciences is proud to bring the outstanding quality of Borosil Type 3.3 Borosilicate glassware to North America for the first time. Manufactured from the same USP Type 1 borosilicate glass that North American scientists have come to trust, Borosil Laboratory Glassware offers unparalleled mechanical strength, clarity, and reliability at competitive prices. And thanks to a unique annealing process, Borosil Glassware is stress-free and incredibly resistant to thermal shock.
Catalog No. 50-154-2839 Supplier Foxx Life Sciences Supplier No. 3083045
From streamlined contours to high-quality glass stopcocks, Borosil Vacuum Desiccators reflect great care and attention to detail. Borosil Desiccators are perfect for drying, and for storage of moisture-sensitive substances. Compatible with any common desiccant. Porcelain plate, ground glass flanges. Rated to 9.6 PSI vacuum when properly cleaned and greased. Avail. from 100mm (XS) to 300mm (XL). Also Avail. in Light-Blocking Amber. In collaboration with Borosil Glass Works, Foxx Life Sciences is proud to bring the outstanding quality of Borosil Type 3.3 Borosilicate glassware to North America for the first time. Manufactured from the same USP Type 1 borosilicate glass that North American scientists have come to trust, Borosil Laboratory Glassware offers unparalleled mechanical strength, clarity, and reliability at competitive prices. And thanks to a unique annealing process, Borosil Glassware is stress-free and incredibly resistant to thermal shock.