Learn More
Synthware™ Vacuum Distillation Apparatus
GREENER_CHOICESDP
Catalog No. 31501042 Shop All Kemtech Products
Change view
Click to view available options
Standard Taper Joint:
10/18, 14/20
10/18, 19/22
14/20, 14/20
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Standard Taper Joint
31-501-042 14/20, 14/20
31-501-040 10/18, 14/20
31-501-041 10/18, 19/22
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 31-501-042 Supplier Synthware™ Supplier No. D101414
Add to Cart
Add to Cart
10/18 joint on side tube to accommodate a 25 mm immersion thermometer.
Specifications
For Use With (Application) Organic Synthesis
Type Vacuum Distillation Apparatus
Material Borosilicate Glass
For Use With (Equipment) 25 mm immersion thermometer
Quantity 1 Ea.
Standard Taper Joint 14/20, 14/20
Joint Type Ground Glass