Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Vacuum Pump Exhaust Filter, no adapter
SDP

Catalog No. 501902693
Encompass

Vacuum pump exhaust filters for vacuum rotary vane pumps remove all visible oil mist and smoke from vacuum pump exhaust, even when it is saturated with oil. The high efficiency filter cartridge continuously drains the collected liquid, allowing recovery of expensive lubricating fluid. Filters are for pumps with flow ratings ranging of 3 cfm in non-hazardous/non-corrosive applications. Port size is 1/2" NPT.

The filter eliminate 99% oil mist and smoke from vacuum pump exhaust and prevents oil accumulation in ductwork. Expensive lubricating oils is automatically returned to pump. Using the filter will eliminate potential OSHA and EPA violations.

Specifications
Maximum Flow Rate: 3 CFM.
Material: Nylon. Maximum Seals: None.
Maximum Temperature: 121 C.
Shipping Pressure: 15PSIG.
Dimensions: 2" Dia. x 3.7" H.
Weight: .25lbs (0.1kg).

Catalog No. 50-190-2693 Supplier Electron Microscopy Sciences Supplier No. 96000
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Vacuum pump exhaust filters for vacuum rotary vane pumps remove all visible oil mist and smoke from vacuum pump exhaust, even when it is saturated with oil. The high efficiency filter cartridge continuously drains the collected liquid, allowing recovery of expensive lubricating fluid. Filters are for pumps with flow ratings ranging of 3 cfm in non-hazardous/non-corrosive applications. Port size is 1/2" NPT.

The filter eliminate 99% oil mist and smoke from vacuum pump exhaust and prevents oil accumulation in ductwork. Expensive lubricating oils is automatically returned to pump. Using the filter will eliminate potential OSHA and EPA violations.

Specifications
Maximum Flow Rate: 3 CFM.
Material: Nylon. Maximum Seals: None.
Maximum Temperature: 121 C.
Shipping Pressure: 15PSIG.
Dimensions: 2" Dia. x 3.7" H.
Weight: .25lbs (0.1kg).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.